Learn More
Grainger Workbench Particleboard 120 W 36 D
Catalog No. 502766595
Encompass_Preferred
The BenchPro Adjustable Height Workbench is a sturdy steel workbench that allows you to adjust the height from 30" to 42" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60" W x 24" D. The bench includes four heavy-duty swivel casters, two with brakes. Weight capacity is 1200 lbs. Assembly required. Backed by a 10-year limited warranty.
Catalog No. 50-276-6595 Supplier Grainger Supplier No. 16Y088
The BenchPro Adjustable Height Workbench is a sturdy steel workbench that allows you to adjust the height from 30" to 42" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60" W x 24" D. The bench includes four heavy-duty swivel casters, two with brakes. Weight capacity is 1200 lbs. Assembly required. Backed by a 10-year limited warranty.