Learn More
Grainger Workbench Particleboard 60 W 30 D
Catalog No. 502766826
Encompass_Preferred
The Grainger 16Y082 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield. It can be used for general purpose applications.
Catalog No. 50-276-6826 Supplier Grainger Supplier No. 16Y082
The Grainger 16Y082 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield. It can be used for general purpose applications.