Learn More
Grainger Workbench Particleboard 72 W 36 D
Catalog No. 502766594
Encompass_Preferred
The BENCHPRO Workbench with Adjustable Height is a heavy-duty steel workbench that allows you to adjust the height from 30" to 42" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60" W x 24" D and can hold up to 1200 lbs. The workbench also features two shelves for storage, four swivel casters for mobility, and a power strip with 4 outlets built into the frame. This height-adjustable workbench provides flexibility for multiple users.
Catalog No. 50-276-6594 Supplier Grainger Supplier No. 16Y086
The BENCHPRO Workbench with Adjustable Height is a heavy-duty steel workbench that allows you to adjust the height from 30" to 42" to accommodate sitting or standing. It has a 1-1/4" thick maple laminate worksurface that measures 60" W x 24" D and can hold up to 1200 lbs. The workbench also features two shelves for storage, four swivel casters for mobility, and a power strip with 4 outlets built into the frame. This height-adjustable workbench provides flexibility for multiple users.