Learn More
Grainger Workbench SS 117 W 30 D
Catalog No. 502765058
Encompass_Preferred
The Grainger Approved Workbench is a sturdy steel workbench that provides a durable and stable work surface. It features an open frame design with four adjustable leveling feet to ensure stability on uneven floors. The workbench has a 1-1/4" thick maple laminate work surface that measures 60" W x 24" D, providing ample workspace. The steel frame and legs have a powder coat finish to prevent rust and corrosion. This industrial-grade workbench can support up to 1200 lbs of distributed weight capacity. Its open base allows for flexible storage options. The Grainger Approved Workbench requires minor assembly and is backed by a 1-year limited warranty.
Catalog No. 50-276-5058 Supplier Grainger Supplier No. 16Y110
The Grainger Approved Workbench is a sturdy steel workbench that provides a durable and stable work surface. It features an open frame design with four adjustable leveling feet to ensure stability on uneven floors. The workbench has a 1-1/4" thick maple laminate work surface that measures 60" W x 24" D, providing ample workspace. The steel frame and legs have a powder coat finish to prevent rust and corrosion. This industrial-grade workbench can support up to 1200 lbs of distributed weight capacity. Its open base allows for flexible storage options. The Grainger Approved Workbench requires minor assembly and is backed by a 1-year limited warranty.