Learn More
Grainger Workbench SS 72 W 30 D
Catalog No. 502767158
Encompass_Preferred
The Workbench SS 72"W x 30"D is a stainless steel workbench that provides a durable and corrosion resistant work surface. It has an open base design with adjustable leveling feet. The workbench features a 1-1/4" thick maple butcher block top that measures 72" wide by 30" deep, providing ample workspace. The height of the workbench is adjustable from 30" up to 37". This industrial grade workbench is made in the USA and can support up to 3000 lbs. Its stainless steel construction makes it ideal for use in wet, high traffic areas. The open base allows for easy cleanability.
Catalog No. 50-276-7158 Supplier Grainger Supplier No. 16Y108
The Workbench SS 72"W x 30"D is a stainless steel workbench that provides a durable and corrosion resistant work surface. It has an open base design with adjustable leveling feet. The workbench features a 1-1/4" thick maple butcher block top that measures 72" wide by 30" deep, providing ample workspace. The height of the workbench is adjustable from 30" up to 37". This industrial grade workbench is made in the USA and can support up to 3000 lbs. Its stainless steel construction makes it ideal for use in wet, high traffic areas. The open base allows for easy cleanability.