Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Koehler™ Bright Star™ WorkSAFE™ Model 2206 LED Flashlight

Catalog No. 19337662
Encompass
Encompass_Preferred
Change view
Click to view available options
Light Source:
LED
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Light Source
19-337-662 LED
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-337-662 Supplier Koehler™ Bright Star™ Supplier No. 08050
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Koehler™ Bright Star WorkSAFE™ Model 2206 LED Flashlight is ideal for use in hazardous environments and is waterproof.

  • Waterproof, IP 67 rating
  • 'D' Cell Alkaline batteries
  • 'Switch Guard' protects switch on impact
  • Glove friendly handle

Certifications

UL Div 1 class 1 Intrinsic Safety

Warranty and Services

Limited lifetime warranty

Specifications

Light Source LED
Lighting Life 30,000 hr.
Weight (English) 5.8 lb.
Weight (Metric) 2.6 kg
Battery Run Time 150 hours
Battery Type 4 x D Cell Alkaline
Height (English) 5.6 in.
Height (Metric) 14.2 cm
Length (English) 8.2 in.
Length (Metric) 20.9 cm
Lumen 90 lm
Width (English) 4.9 in.
Width (Metric) 12.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.