Learn More
MSC Value Collection Modular Workstation, Plastic Laminate Top, 5 Drawers, 5,400 lb Capacity, 1 ea.
Catalog No. 5000511071
Encompass
This modular workstation is designed for heavy-duty storage and workspace applications. Featuring a durable steel frame and a plastic laminate top, it provides a stable and reliable surface for various tasks. The workstation includes five drawers for organized storage and is engineered to support significant loads.
  • Features a high-capacity design of 5,400 lb
  • Equipped with 5 integrated drawers for storage
  • Constructed with a sturdy steel body
  • Equipped with a durable plastic laminate top
  • Designed with a comfort edge for improved ergonomics
  • Dimensions: 72 in. wide by 30 in. deep by 32 in. high
  • Top thickness of 1-3/4 in.
Catalog No. 50-005-11071 Supplier MSC Supplier No. 91683003
This modular workstation is designed for heavy-duty storage and workspace applications. Featuring a durable steel frame and a plastic laminate top, it provides a stable and reliable surface for various tasks. The workstation includes five drawers for organized storage and is engineered to support significant loads.
  • Features a high-capacity design of 5,400 lb
  • Equipped with 5 integrated drawers for storage
  • Constructed with a sturdy steel body
  • Equipped with a durable plastic laminate top
  • Designed with a comfort edge for improved ergonomics
  • Dimensions: 72 in. wide by 30 in. deep by 32 in. high
  • Top thickness of 1-3/4 in.