Learn More
MSC WorkSmart S-Hook, Low Carbon Steel, Electrogalvanized, 5/32 in Opening, 16 Lb Capacity
Catalog No. 5000533027
Encompass
This S-hook is constructed from low carbon steel with an electrogalvanized finish, offering a durable solution for various applications. It features a 5/32 in opening and a 16 Lb capacity, suitable for both marine and industrial environments.
  • Constructed from low carbon steel
  • Features an electrogalvanized zinc finish
  • Designed with a 5/32 in opening
  • Offers a 16 Lb load capacity
  • Wire size measures 0.105 in
  • Overall length of 1 in
  • Inner diameter of 5/16 in
  • Ideal for marine and industrial applications
Catalog No. 50-005-33027 Supplier MSC Supplier No. 13574801
This S-hook is constructed from low carbon steel with an electrogalvanized finish, offering a durable solution for various applications. It features a 5/32 in opening and a 16 Lb capacity, suitable for both marine and industrial environments.
  • Constructed from low carbon steel
  • Features an electrogalvanized zinc finish
  • Designed with a 5/32 in opening
  • Offers a 16 Lb load capacity
  • Wire size measures 0.105 in
  • Overall length of 1 in
  • Inner diameter of 5/16 in
  • Ideal for marine and industrial applications