Learn More
MSC Value Collection Industrial Work Bench, Steel and Hardboard, 60 in. x 30 in., 2,500 lb. Capacity, 1 ea.
Catalog No. 5000515293
Encompass
This industrial work bench provides a robust and reliable surface for demanding tasks in manufacturing and maintenance environments. Designed for stability and durability, it features a spacious work area and a high load capacity, making it suitable for a wide range of industrial applications.
  • Load capacity of 2,500 lb
  • Overall dimensions are 60 in. wide by 30 in. deep by 34-5/8 in. high
  • Constructed with a steel and hardboard top
  • Features a 1-5/8 in. thick top
  • Equipped with adjustable legs
  • Finished in gray
Catalog No. 50-005-15293 Supplier MSC Supplier No. 91683508
This industrial work bench provides a robust and reliable surface for demanding tasks in manufacturing and maintenance environments. Designed for stability and durability, it features a spacious work area and a high load capacity, making it suitable for a wide range of industrial applications.
  • Load capacity of 2,500 lb
  • Overall dimensions are 60 in. wide by 30 in. deep by 34-5/8 in. high
  • Constructed with a steel and hardboard top
  • Features a 1-5/8 in. thick top
  • Equipped with adjustable legs
  • Finished in gray