Learn More
Grainger Pro-Line Open Frame Workstand, Steel, 48 in. x 24 in., 500 lb. Capacity, 1 ea
Catalog No. 502768108
Encompass_Preferred
This open frame workstand provides a sturdy and versatile workspace solution for industrial or laboratory applications. Designed for reliability and durability, it features a heavy-duty steel construction suitable for demanding environments.
  • Overall dimensions of 48 in. width by 24 in. depth by 68 3/4 in. height
  • Load capacity of 500 lb.
  • Constructed from 16 ga steel
  • Beige finish
  • Includes mounting hardware
  • Ships unassembled
Catalog No. 50-276-8108 Supplier Grainger Supplier No. 16Y938
This open frame workstand provides a sturdy and versatile workspace solution for industrial or laboratory applications. Designed for reliability and durability, it features a heavy-duty steel construction suitable for demanding environments.
  • Overall dimensions of 48 in. width by 24 in. depth by 68 3/4 in. height
  • Load capacity of 500 lb.
  • Constructed from 16 ga steel
  • Beige finish
  • Includes mounting hardware
  • Ships unassembled