Learn More
Cole-Parmer Argos Technologies PolarSafe Cryovial Rack, 40, Red; 1/Ea
Catalog No. 501960638 Shop All Cole-Parmer Products
Encompass_Preferred
This rack securely holds up to 40 self-standing cryogenic vials. It features a unique locking design of wells that also allows for one-handed opening and closing of caps.
Catalog No. 50-196-0638 Supplier Cole-Parmer Supplier No. 0439572
This rack securely holds up to 40 self-standing cryogenic vials. It features a unique locking design of wells that also allows for one-handed opening and closing of caps.