Learn More
Analytical Sales & Services Workstation Dispensing Bin - One Bin
Catalog No. 5000805662
Encompass
Workstation Dispensing Bin - One Bin 184W x 194H x 232mmD Know inventory levels of pipette tips, tubes, fittings, finger cots and other bulk items at a glance. Fliptop covers on generous openings make filling and dispensing easy and keep products dust-free. With non-skid rubber feet.
Catalog No. 50-008-05662 Supplier Analytical Sales & Services Supplier No. 97321
Workstation Dispensing Bin - One Bin 184W x 194H x 232mmD Know inventory levels of pipette tips, tubes, fittings, finger cots and other bulk items at a glance. Fliptop covers on generous openings make filling and dispensing easy and keep products dust-free. With non-skid rubber feet.