Learn More
Analytical Sales & Services Workstation Dispensing Bins - Three Bins
Catalog No. 5000805927
Encompass
Workstation Dispensing Bins - Three Bins 217W x 170H x 217mmD Know inventory levels of pipette tips, tubes, fittings, finger cots and other bulk items at a glance. Fliptop covers on generous openings make filling and dispensing easy and keep products dust-free. With non-skid rubber feet.
Catalog No. 50-008-05927 Supplier Analytical Sales & Services Supplier No. 97323
Workstation Dispensing Bins - Three Bins 217W x 170H x 217mmD Know inventory levels of pipette tips, tubes, fittings, finger cots and other bulk items at a glance. Fliptop covers on generous openings make filling and dispensing easy and keep products dust-free. With non-skid rubber feet.