Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Workstation Laboratory Identification Software Suite E-media

Catalog No. 19129182 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Brady Label Printers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-129-182 Brady Label Printers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-129-182 Supplier Brady™ Supplier No. BWSLABSEM

missing translation for 'validMainframeMsg'

Brady™ Workstation Laboratory Identification Software Suite E-media opens up more options in a lab and allows you to create custom labels.

  • For use with: BBP™85 Industrial Sign and Label Printer, BBP™81 Label Printer, BBP™35 Multi-Color Sign and Label Printer, BBP™33 Label Printer, BBP™12 Label Printer, BBP™11 Label Printer, BMP™71 Label Printer, BMP™61 Labeling System, BMP™51 Label Printer, BMP™41 Label Printer, BMP™53 Label Printer, BradyJet™ J5000, BradyPrinter i7100, BradyPrinter i5100, BRADY IP™ Series Label Printer, LABXPERT™ Handheld Label Maker, PR300 Plus Industrial Label Printer, PR600 Plus Industrial Label Printer, GlobalMark™ 2 Color and Cut Industrial Label Maker, GlobalMark™ Multicolor Industrial Label Maker, IDXPERT™ Handheld Label Maker, BradyJet™ J2000, J5000 Color Label Printer, BradyPrinter S3100, TLS 2200™ Thermal Labeling System, BBP™31 Sign and Label Printer, BBP™37 Color & Cut Sign and Label Printer, BradyPrinter™ M611
  • Languages: Bulgarian, Chinese (Simplified), Croatian, Czech, Danish, Dutch, English (US), Estonian, Finnish, French (France), German, Hungarian, Italian, Japanese, Korean, Spanish (Mexico), Norwegian, Polish, Portuguese (Portugal), Romanian, Russian, Slovak, Slovenian, Spanish (Spain), Swedish, Chinese (Traditional), Turkish
  • Operating Systems: Windows™ 10, Windows™ 7 (32- or 64-bit), Windows™ 7 Enterprise, Windows™ 7 Professional, Windows™ 7 Ultimate, Windows™ 8 (32- or 64-bit), Windows™ 8 Professional (32 or 64 bit), Windows™ 8.1 (32- or 64-bit), Windows™ 8.1 Enterprise, Windows™ 8.1 Professional
  • Design custom labels that include text, barcodes, graphics and shapes
  • Create sequences with numeric or alphabetic sequential text or barcodes
  • Import data and add information from Excel, text files or databases
  • Make templates with locked and editable elements for consistent label creation
  • Save time by grouping multiple label files and print all at once
  • Advanced Sequence - Create dependent and complex sequences for label creation
  • Batch Print - Efficiently group and print multiple files at once
  • Print Partner - Quickly create labels by filling in a template and printing

Specifications

Product Type Software
For Use With (Equipment) Brady Label Printers
Width (English) 0.002 in.
Resolution 1024 x 768
Software Type Identification Label Printing
For Use With (Application) Laboratory identification
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.