Learn More
Description
Requires
Internet connection
- Print Partner has no design capabilities, so it is deployable on multiple locations where only printing is required
- Users can only complete designated fields so you are always sure that your labels meet the specifications of you or your customers
- Runs as an app within the Brady™ Workstation platform or as a stand-alone application
- The simplified user interface only gives you the required functionality while limiting training for users
- Windows™ 7 Professional; Windows™ 7 Ultimate; Windows™ 7 Enterprise; Windows™ 8.1 Enterprise; Windows™ 8.1 Professional
- Single user license
Specifications
Specifications
| Product Type | Software |
| For Use With (Equipment) | BBP11 Label Printer, BBP33 Label Printer, Brady 1244, Brady 1344, Brady IP Series Label Printer, BradyPrinter 300PR+, Brady 2461/3481/6441 Series, Brady MVP Plus Series, Brady X Plus II Series Label Machine, BMP41 Label Printer, BMP71 Label Printer, Laser Printers, Ink Jet Printers, BMP53 Label Printer, BMP51 Label Printer |
| Width (English) | 0.002 in. |
| Format | E-Media |
| Memory | 200 MB, 1.5 Gigahertz (GHZ) Processor or Faster with 2 GB RAM |
| For Use With (Application) | Label printing |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.