Learn More
Brady™ Workstation Scan and Print Application Email Activation Code
Catalog No. 19129128 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-129-128 1 Ea.
1 options
Catalog No. 19-129-128 Supplier Brady™ Supplier No. BWSSPEM
Brady Workstation Scan and Print Application Email Activation Code, create label workflows to populate your own custom label templates.
Scan Mode - Scan existing barcodes to capture data and transfer onto a new label template.CVS Mode - Scan the name of a CSV file to print a specified batch of labels. Custom Script Mode - Perform advanced label operations such as parse scanned data or serialize labels. Scalable Workflows - Expand workflows to additional workstations on large projects using the Export and Import feature. Setup as many workflows as you need.
Specifications
For Use With (Equipment) Brady Printers, Brady Scanners
Width (Metric) N/A
Includes Email Activation Code for Software
Operating System Windows™ 7 (32- or 64-bit);Windows™ 7 Enterprise;Windows™ 7 Professional;Windows™ 7 SPI;Windows™ 7 Ultimate;Windows™ 8 (32- or 64-bit);Windows™ 8 Professional (32 or 64 bit);Windows™ 8.1 (32- or 64-bit);Windows™ 8.1 Enterprise;Windows™ 8.1 Professional;Windows™ 10
Quantity 1 Ea.