Learn More
Enterprise Technology Solutions World Centric Takeout Containers,Hinged,PLA,BioBased,PlantSugar,FreezerSafe,f/Cold-WarmFood,8"x8"x3",300/CT,Clear
SDP
Catalog No. 5000789807
Encompass_Preferred
World Centric Compostable Clear Hinged Containers are made from PLA, a bio-based plastic made from plant sugars. Great for takeout restaurants, cafes and grocery stores, these eco-friendly takeout containers are light-weight and freezer-safe. PLA products are suitable for cold to warm foods, and should be stored in a cool place (below 110-degrees Fahrenheit and out of direct sunlight). They're USDA Certified Biobased, Compost Manufacturing Alliance (CMA) Certified Compostable, NSF Industrial Certified Compostable and TUV Austria OK Industrial Certified Compostable. As a certified B Corporation, World Centric is dedicated to upholding the highest standards of social and environmental responsibility, transparency, and accountability. We proudly give 25-percent of our profits to grassroots organizations driving global change. Choose World Centric compostable foodservice products and make a difference today.
Catalog No. 50-007-89807 Supplier Enterprise Technology Solutions Supplier No. WORKLCS8N
World Centric Compostable Clear Hinged Containers are made from PLA, a bio-based plastic made from plant sugars. Great for takeout restaurants, cafes and grocery stores, these eco-friendly takeout containers are light-weight and freezer-safe. PLA products are suitable for cold to warm foods, and should be stored in a cool place (below 110-degrees Fahrenheit and out of direct sunlight). They're USDA Certified Biobased, Compost Manufacturing Alliance (CMA) Certified Compostable, NSF Industrial Certified Compostable and TUV Austria OK Industrial Certified Compostable. As a certified B Corporation, World Centric is dedicated to upholding the highest standards of social and environmental responsibility, transparency, and accountability. We proudly give 25-percent of our profits to grassroots organizations driving global change. Choose World Centric compostable foodservice products and make a difference today.