This jacket offers high-level cut protection and easy on/off comfort with a full zip design. It is made from extremely strong, high-performance fibers, providing significant cut and abrasion resistance in high-risk zones.
- Full-length zipper closure for easy on/off
- Reinforced belly, neck, and forearms for significant cut and abrasion resistance
- Knitted mesh back reduces overall garment weight and releases body heat
- Comfort Collar (4 in) has a cooling effect on the skin and provides ANSI A7 protection for the neck
- Thumb holes keep sleeves from riding up
- Easy to wash in cold water; tumble dry on low heat
- Reinforced knit across the abdomen rated ANSI A6
- Forearms score ANSI A5 for cut protection
- Upper chest, biceps, and back panel deliver ANSI A4 cut resistance
- Intentionally long to offer protection to the groin