Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Worldwide Glass Resource 13-425 Polypropylene Screw Thread Closures

Catalog No. 14812128 Shop All Worldwide Glass Resource Products
Change view
Click to view available options
Pre-slit:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit
14-812-128 Yes
14-812-127 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-812-128 Supplier Worldwide Glass Resource Supplier No. 0016168
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Open-top closures for chromatography vials

  • 13-425 screw thread finish
  • Polypropylene construction

Specifications

Color Black
Material Polypropylene
Pre-slit Yes
Closure Size 13-425
For Use With (Equipment) Chromatography Vials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.