Learn More
Worldwide Glass Resource 8-425 Polypropylene Screw Thread Closures
Catalog No. 1481240 Shop All Worldwide Glass Resource Products
Change view
Click to view available options
Color:
Black
Catalog No. Color
14-812-40 Black
1 options
Catalog No. 14-812-40 Supplier Worldwide Glass Resource Supplier No. 0016001
Add to Cart
Add to Cart
Open-top closures for use with chromatography vials
  • Polypropylene construction
  • 8-425 screw thread finish
  • Black with red PTFE/white silicone
  • Sold in case of 1000
Specifications
Color Black
Material Polypropylene
Closure Size 8-425