Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Worm Composter Classroom Kit
SDPSCI_ED

Catalog No. S37285 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Quantity:
14 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S37285 14 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog No. S37285 Supplier Carolina™ Supplier No. 141560

This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-766-7000. Provide the NEED BY DATE to the customer service agent.

missing translation for 'validMainframeMsg'

There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Introduces students to the importance of decomposers in the food web. Carolina™ Biological Supply Worm Composter Classroom Kit explains the concept of recycling through composting.

  • Grade: K and up
  • Observe quick and efficient conversion of organic matter into enriched soil components by redworms once kitchen waste is added
  • Composting container is constructed of recycled plastic
  • Has 3 vents for proper air flow
  • Kit includes worm composter with lid, red worms (500), worm bedding, and the classroom activity book Worms Eat Our Garbage
  • Dimensions: 43.1 x 55.8 x 30.4cm (17 x 22 x 12 in.)

Specifications

For Use With (Application) Recycling compost
Quantity 14 gal.
Includes Worm composter with lid, red worms (500), worm bedding, and the classroom activity book Worms Eat Our Garbage
Sufficient For Class of 30 students
Product Type Worm Composter Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.