Learn More
McMaster-Carr Worm-Drive Clamps for Firm Hose and Tube with 410 Stainless Steel Screw, 1/2" Band Width, 1/2" to 29/32" Clamp ID, 10/pk
Catalog No. NC1148688
Encompass
So easy to install, it's no wonder these are the most frequently used style of hose clamps. Clamps are for firm plastic and rubber hose and tube. Do not exceed the maximum torque or clamps may be damaged.
Catalog No. NC1148688 Supplier McMaster-Carr Supplier No. 5416K32
So easy to install, it's no wonder these are the most frequently used style of hose clamps. Clamps are for firm plastic and rubber hose and tube. Do not exceed the maximum torque or clamps may be damaged.