Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 1/2 ID PK10
Catalog No. 502765293
Encompass_Preferred
The Worm Gear Hose Clamp 1/2" W, 1-1/2" ID from Grainger is a worm gear style hose clamp with a band width of 1/2" and an internal diameter of 1-1/2". It is made from 201 stainless steel for corrosion resistance and has a zinc plated band and screw housing for additional protection. The clamp has a 360 degree tightening mechanism for even distribution of clamping force. It is sold in a pack of 10 clamps. The clamps are designed for automotive and industrial hose connections.
Catalog No. 50-276-5293 Supplier Grainger Supplier No. 16P283
The Worm Gear Hose Clamp 1/2" W, 1-1/2" ID from Grainger is a worm gear style hose clamp with a band width of 1/2" and an internal diameter of 1-1/2". It is made from 201 stainless steel for corrosion resistance and has a zinc plated band and screw housing for additional protection. The clamp has a 360 degree tightening mechanism for even distribution of clamping force. It is sold in a pack of 10 clamps. The clamps are designed for automotive and industrial hose connections.