Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 1/4 ID PK10
Catalog No. 502767302
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1/4 to 1-1/4 inches. This hose clamp features a rolled edge, screw housing and worm drive adjusting screw. It is designed for automotive and industrial hose connections. The 201 stainless steel offers good corrosion resistance. This is a high quality worm gear hose clamp suitable for a wide range of hose clamping applications.
Catalog No. 50-276-7302 Supplier Grainger Supplier No. 16P282
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1/4 to 1-1/4 inches. This hose clamp features a rolled edge, screw housing and worm drive adjusting screw. It is designed for automotive and industrial hose connections. The 201 stainless steel offers good corrosion resistance. This is a high quality worm gear hose clamp suitable for a wide range of hose clamping applications.