Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 1/2 ID PK10
Catalog No. 502767669
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/4 inch. This hose clamp features a rolled edge, screw housing and worm drive screw to provide constant tension. It is designed for automotive and industrial hose connections. The 201 stainless steel offers good corrosion resistance. This worm gear clamp meets DIN 3017 specifications.
Catalog No. 50-276-7669 Supplier Grainger Supplier No. 16P305
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/4 inch. This hose clamp features a rolled edge, screw housing and worm drive screw to provide constant tension. It is designed for automotive and industrial hose connections. The 201 stainless steel offers good corrosion resistance. This worm gear clamp meets DIN 3017 specifications.