Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 1/4 ID PK10
Catalog No. 502765973
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel is a 304 stainless steel worm gear clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and features a 360 degree tightening mechanism. The clamp has a temperature range of -100 to 1000 degrees Fahrenheit. This hose clamp is made in the USA and is RoHS compliant. The clamp meets ASME B16.20 and SAE J1508 standards. It can be used for automotive, industrial, and marine applications. The clamp comes in sizes ranging from 1/2 inch to 4 inches.
Catalog No. 50-276-5973 Supplier Grainger Supplier No. 16P304
The Worm Gear Hose Clamp 304 Stainless Steel is a 304 stainless steel worm gear clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and features a 360 degree tightening mechanism. The clamp has a temperature range of -100 to 1000 degrees Fahrenheit. This hose clamp is made in the USA and is RoHS compliant. The clamp meets ASME B16.20 and SAE J1508 standards. It can be used for automotive, industrial, and marine applications. The clamp comes in sizes ranging from 1/2 inch to 4 inches.