Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 3/4 ID PK10
Catalog No. 502767365
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1/4 to 1-1/4 inches. This hose clamp is designed for automotive and industrial applications. The 201 stainless steel offers good corrosion resistance. The worm drive allows the clamp to be tightened by turning the screw to provide a tight, secure hold. This hose clamp meets DIN 3017 specifications.
Catalog No. 50-276-7365 Supplier Grainger Supplier No. 16P284
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1/4 to 1-1/4 inches. This hose clamp is designed for automotive and industrial applications. The 201 stainless steel offers good corrosion resistance. The worm drive allows the clamp to be tightened by turning the screw to provide a tight, secure hold. This hose clamp meets DIN 3017 specifications.