Learn More
Grainger Worm Gear Hose Clamp 1/2 W 1 ID PK10
Catalog No. 502765157
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1-1/4 inches to 2-1/2 inches. This hose clamp features a rolled edge, screw housing and worm drive screw to tighten the band. It can provide up to 750 pounds of clamping force and is designed for automotive and industrial hose connections. The 201 stainless steel offers good corrosion resistance. This is a high quality worm gear hose clamp suitable for a wide range of hose clamping applications.
Catalog No. 50-276-5157 Supplier Grainger Supplier No. 16P277
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1-1/4 inches to 2-1/2 inches. This hose clamp features a rolled edge, screw housing and worm drive screw to tighten the band. It can provide up to 750 pounds of clamping force and is designed for automotive and industrial hose connections. The 201 stainless steel offers good corrosion resistance. This is a high quality worm gear hose clamp suitable for a wide range of hose clamping applications.