Learn More
Grainger Worm Gear Hose Clamp 1/2 W 3/4 ID PK10
Catalog No. 502765937
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1-1/4 inches to 2-1/4 inches. This hose clamp features a 201 stainless steel band and housing and a worm drive screw of hardened steel. It is designed for automotive and industrial hose connections. The clamp provides constant tension and heavy duty performance. This is an OE approved hose clamp.
Catalog No. 50-276-5937 Supplier Grainger Supplier No. 16P276
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1-1/4 inches to 2-1/4 inches. This hose clamp features a 201 stainless steel band and housing and a worm drive screw of hardened steel. It is designed for automotive and industrial hose connections. The clamp provides constant tension and heavy duty performance. This is an OE approved hose clamp.