Learn More
Grainger Worm Gear Hose Clamp 1/2 W 3/4 ID PK10
Catalog No. 502767261
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm gear style hose clamp made from 201 stainless steel banding. It has a band width of 7/16", a clamping range of 5/8" to 1-1/4", and a screw housing length of 13/16". The 201 stainless steel offers good corrosion resistance. This hose clamp is designed for automotive and industrial applications. The worm gear mechanism allows for easy tightening.
Catalog No. 50-276-7261 Supplier Grainger Supplier No. 16P302
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm gear style hose clamp made from 201 stainless steel banding. It has a band width of 7/16", a clamping range of 5/8" to 1-1/4", and a screw housing length of 13/16". The 201 stainless steel offers good corrosion resistance. This hose clamp is designed for automotive and industrial applications. The worm gear mechanism allows for easy tightening.