Learn More
Grainger Worm Gear Hose Clamp 1/2 W 3/4 ID PK10
Catalog No. 502767386
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel from Grainger is a worm gear style hose clamp made from 201 stainless steel. It has a band width of 1/2" and a clamping range of 1/4" to 1-1/4". The clamp features a 360 degree tightening mechanism with screw take-up on the band. It is designed for automotive and industrial hose connections. This stainless steel hose clamp provides good corrosion resistance and can be used for exhaust system connections.
Catalog No. 50-276-7386 Supplier Grainger Supplier No. 16P280
The Worm Gear Hose Clamp 201 Stainless Steel from Grainger is a worm gear style hose clamp made from 201 stainless steel. It has a band width of 1/2" and a clamping range of 1/4" to 1-1/4". The clamp features a 360 degree tightening mechanism with screw take-up on the band. It is designed for automotive and industrial hose connections. This stainless steel hose clamp provides good corrosion resistance and can be used for exhaust system connections.