Learn More
Grainger Worm Gear Hose Clamp 1/2 W 4 ID PK10
Catalog No. 502765974
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel from Grainger is a worm gear style hose clamp made from 201 stainless steel. It has a band width of 1/2 inch and a clamping range of 1/4 inch to 1-1/4 inches. The clamp features a rolled edge and screw housing, and it is designed for automotive and industrial hose connections. This hose clamp provides up to 1550 psi working pressure. It meets SAE J1508 Type 1 performance requirements. The clamp's stainless steel construction resists corrosion and has a matte finish.
Catalog No. 50-276-5974 Supplier Grainger Supplier No. 16P312
The Worm Gear Hose Clamp 201 Stainless Steel from Grainger is a worm gear style hose clamp made from 201 stainless steel. It has a band width of 1/2 inch and a clamping range of 1/4 inch to 1-1/4 inches. The clamp features a rolled edge and screw housing, and it is designed for automotive and industrial hose connections. This hose clamp provides up to 1550 psi working pressure. It meets SAE J1508 Type 1 performance requirements. The clamp's stainless steel construction resists corrosion and has a matte finish.