Learn More
Grainger Worm Gear Hose Clamp 5/8 W 1 ID PK10
Catalog No. 502763981
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel by Oetiker is a 304 stainless steel worm gear clamp with a band width of 7/16 inch. It has a worm screw drive for tightening and can withstand pressures up to 350 psi. The clamp has a temperature range of -100 to 500 degrees Fahrenheit. This hose clamp is made in Switzerland and conforms to DIN 3017 standards. The clamp has a tensile strength of 100,000 psi minimum.
Catalog No. 50-276-3981 Supplier Grainger Supplier No. 16P234
The Worm Gear Hose Clamp 304 Stainless Steel by Oetiker is a 304 stainless steel worm gear clamp with a band width of 7/16 inch. It has a worm screw drive for tightening and can withstand pressures up to 350 psi. The clamp has a temperature range of -100 to 500 degrees Fahrenheit. This hose clamp is made in Switzerland and conforms to DIN 3017 standards. The clamp has a tensile strength of 100,000 psi minimum.