Learn More
Grainger Worm Gear Hose Clamp 5/8 W 2 3/4 ID PK10
Catalog No. 502763971
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel by Oetiker is a 304 stainless steel worm gear hose clamp with a band width of 15/16 in. It has a clamping range of 1-1/4 in. to 2-1/4 in. This hose clamp features a rolled edge and raised screw housing to prevent hose damage. The worm gear mechanism allows for easy tightening. This stainless steel hose clamp is corrosion resistant and provides reliable performance in heavy duty applications. The clamp meets DIN 3017 standards.
Catalog No. 50-276-3971 Supplier Grainger Supplier No. 16P238
The Worm Gear Hose Clamp 304 Stainless Steel by Oetiker is a 304 stainless steel worm gear hose clamp with a band width of 15/16 in. It has a clamping range of 1-1/4 in. to 2-1/4 in. This hose clamp features a rolled edge and raised screw housing to prevent hose damage. The worm gear mechanism allows for easy tightening. This stainless steel hose clamp is corrosion resistant and provides reliable performance in heavy duty applications. The clamp meets DIN 3017 standards.