Learn More
Grainger Worm Gear Hose Clamp 5/8 W 3 3/4 ID PK10
Catalog No. 502764024
Encompass_Preferred
The Worm Gear Hose Clamp is a stainless steel worm drive hose clamp with a band width of 7/8 in. It has a clamping range of 1-1/4 in. to 2 in. The clamp features a 360 degree tightening mechanism with screw take-up on the band. The band is made of 201 stainless steel for strength and corrosion resistance. The worm drive has an 80 teeth per inch gear ratio. This heavy duty hose clamp is commonly used for industrial and automotive applications. The clamp meets SAE J1508 Type 1 standards.
Catalog No. 50-276-4024 Supplier Grainger Supplier No. 16P224
The Worm Gear Hose Clamp is a stainless steel worm drive hose clamp with a band width of 7/8 in. It has a clamping range of 1-1/4 in. to 2 in. The clamp features a 360 degree tightening mechanism with screw take-up on the band. The band is made of 201 stainless steel for strength and corrosion resistance. The worm drive has an 80 teeth per inch gear ratio. This heavy duty hose clamp is commonly used for industrial and automotive applications. The clamp meets SAE J1508 Type 1 standards.