Learn More
Grainger Worm Gear Hose Clamp 5/8 W 5 1/4 ID PK10
Catalog No. 502763982
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel from Grainger is a 304 stainless steel worm gear hose clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and features a 360 degree tightening mechanism. The clamp has a working range of 5/8 inch to 1-1/4 inch diameter. It is made in the USA and conforms to ASME B16.20 and SAE J1508 standards. The clamp is available in a package quantity of 10 pieces.
Catalog No. 50-276-3982 Supplier Grainger Supplier No. 16P243
The Worm Gear Hose Clamp 304 Stainless Steel from Grainger is a 304 stainless steel worm gear hose clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and features a 360 degree tightening mechanism. The clamp has a working range of 5/8 inch to 1-1/4 inch diameter. It is made in the USA and conforms to ASME B16.20 and SAE J1508 standards. The clamp is available in a package quantity of 10 pieces.