Learn More
Grainger Worm Gear Hose Clamp 5/8 W 5 3/4 ID PK10
Catalog No. 502764629
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel by Oetiker is a 304 stainless steel worm gear hose clamp with a band width of 7/16 inches. It has a worm screw drive for tightening and can withstand pressures up to 350 psi. The clamp has a temperature range of -100 to 500 degrees Fahrenheit. This heavy duty hose clamp is recommended for automotive, industrial, and hydraulic applications. The clamp meets DIN 3017 standards and is made in the USA.
Catalog No. 50-276-4629 Supplier Grainger Supplier No. 16P244
The Worm Gear Hose Clamp 304 Stainless Steel by Oetiker is a 304 stainless steel worm gear hose clamp with a band width of 7/16 inches. It has a worm screw drive for tightening and can withstand pressures up to 350 psi. The clamp has a temperature range of -100 to 500 degrees Fahrenheit. This heavy duty hose clamp is recommended for automotive, industrial, and hydraulic applications. The clamp meets DIN 3017 standards and is made in the USA.