Learn More
Grainger Worm Gear Hose Clamp 5/8 W 5 3/4 ID PK10
Catalog No. 502763897
Encompass_Preferred
The Grainger worm gear tube hose clamps have a maximum inside diameter of 12 inches. They are made of stainless steel for corrosion resistance and have a 360 degree tightening mechanism for secure clamping. The clamps feature a rolled edge to prevent damage to the hose and have a screw housing to protect the adjusting screw. They can accommodate a wide range of hose sizes and materials. The clamps are available in various band widths and screw sizes to provide the proper tightening force. Overall, these are heavy-duty, adjustable clamps well-suited for industrial hose and tube connections.
Catalog No. 50-276-3897 Supplier Grainger Supplier No. 16P228
The Grainger worm gear tube hose clamps have a maximum inside diameter of 12 inches. They are made of stainless steel for corrosion resistance and have a 360 degree tightening mechanism for secure clamping. The clamps feature a rolled edge to prevent damage to the hose and have a screw housing to protect the adjusting screw. They can accommodate a wide range of hose sizes and materials. The clamps are available in various band widths and screw sizes to provide the proper tightening force. Overall, these are heavy-duty, adjustable clamps well-suited for industrial hose and tube connections.