Learn More
Grainger Worm Gear Hose Clamp 5/8 W 6 3/4 ID PK10
Catalog No. 502763989
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel from Grainger is a 304 stainless steel worm gear clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and features a 360 degree tightening mechanism. The clamp has a temperature range of -65 to 300 degrees Fahrenheit. This hose clamp is made in the USA and is RoHS compliant. The clamp meets SAE J1508 Type 1 performance requirements.
Catalog No. 50-276-3989 Supplier Grainger Supplier No. 16P246
The Worm Gear Hose Clamp 304 Stainless Steel from Grainger is a 304 stainless steel worm gear clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and features a 360 degree tightening mechanism. The clamp has a temperature range of -65 to 300 degrees Fahrenheit. This hose clamp is made in the USA and is RoHS compliant. The clamp meets SAE J1508 Type 1 performance requirements.