Learn More
Grainger Worm Gear Hose Clamp 5/8 W 6 3/4 ID PK10
Catalog No. 502763709
Encompass_Preferred
The Worm Gear Hose Clamp is a stainless steel hose clamp with a worm gear tightening mechanism. It has a band width of 1/2 inch and a clamping range of 5/8 inch to 1-1/4 inches. The clamp is made from 201 stainless steel for corrosion resistance. The worm gear allows for tool-free tightening by hand. This hose clamp is ideal for automotive, industrial, and marine applications. The clamp meets SAE J1508 Type 1 standards.
Catalog No. 50-276-3709 Supplier Grainger Supplier No. 16P230
The Worm Gear Hose Clamp is a stainless steel hose clamp with a worm gear tightening mechanism. It has a band width of 1/2 inch and a clamping range of 5/8 inch to 1-1/4 inches. The clamp is made from 201 stainless steel for corrosion resistance. The worm gear allows for tool-free tightening by hand. This hose clamp is ideal for automotive, industrial, and marine applications. The clamp meets SAE J1508 Type 1 standards.