Learn More
Grainger Worm Gear Hose Clamp 5/8 W 7 1/4 ID PK10
Catalog No. 502764628
Encompass_Preferred
The Worm Gear Hose Clamp 304 Stainless Steel from Grainger is a worm gear style hose clamp made from 304 stainless steel. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/4 inch. The clamp features a 360 degree tightening mechanism with screw take-up on the band. It is designed for automotive and industrial hose connections. This stainless steel worm gear clamp provides corrosion resistance and a tight, heavy duty grip.
Catalog No. 50-276-4628 Supplier Grainger Supplier No. 16P231
The Worm Gear Hose Clamp 304 Stainless Steel from Grainger is a worm gear style hose clamp made from 304 stainless steel. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/4 inch. The clamp features a 360 degree tightening mechanism with screw take-up on the band. It is designed for automotive and industrial hose connections. This stainless steel worm gear clamp provides corrosion resistance and a tight, heavy duty grip.