Learn More
Grainger Worm Gear Hose Clamp 5/8 W 8 1/4 ID PK10
Catalog No. 502765265
Encompass_Preferred
The Grainger 16P249 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8/1.2 amps at full/locked rotor load. This electric motor is designed for general purpose applications.
Catalog No. 50-276-5265 Supplier Grainger Supplier No. 16P249
The Grainger 16P249 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8/1.2 amps at full/locked rotor load. This electric motor is designed for general purpose applications.