Learn More
Grainger Worm Gear Hose Clamp 5/8 W 8 1/4 ID PK10
Catalog No. 502763713
Encompass_Preferred
The Worm Gear Hose Clamp 5/8" W, 8-1/4" ID from Grainger is a worm gear style hose clamp with a band width of 5/8 inches and an internal diameter range of 8-1/4 inches. It is constructed from 201 stainless steel for corrosion resistance and has a tensile strength of 30,000 psi. The clamp features a 360 degree tightening mechanism with screw housing to distribute clamping force evenly for secure connections. It can accommodate a wide range of hose sizes and is sold in packs of 10 clamps. This heavy-duty hose clamp provides a reliable, adjustable connection for industrial and automotive applications.
Catalog No. 50-276-3713 Supplier Grainger Supplier No. 16P233
The Worm Gear Hose Clamp 5/8" W, 8-1/4" ID from Grainger is a worm gear style hose clamp with a band width of 5/8 inches and an internal diameter range of 8-1/4 inches. It is constructed from 201 stainless steel for corrosion resistance and has a tensile strength of 30,000 psi. The clamp features a 360 degree tightening mechanism with screw housing to distribute clamping force evenly for secure connections. It can accommodate a wide range of hose sizes and is sold in packs of 10 clamps. This heavy-duty hose clamp provides a reliable, adjustable connection for industrial and automotive applications.