Learn More
Grainger Worm Gear Hose Clamp 9/16 W 1 ID PK10
Catalog No. 502767684
Encompass_Preferred
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/2 inch. This hose clamp features a rolled edge, screw housing, and worm drive screw. It provides constant tension and heavy duty performance. The 201 stainless steel offers good corrosion resistance. This worm gear clamp is designed for automotive and industrial hose connections.
Catalog No. 50-276-7684 Supplier Grainger Supplier No. 16P250
The Worm Gear Hose Clamp 201 Stainless Steel by Oetiker is a worm drive hose clamp made from 201 stainless steel banding. It has a band width of 1/2 inch and a clamping range of 1-1/4 inch to 2-1/2 inch. This hose clamp features a rolled edge, screw housing, and worm drive screw. It provides constant tension and heavy duty performance. The 201 stainless steel offers good corrosion resistance. This worm gear clamp is designed for automotive and industrial hose connections.