Learn More
Grainger Worm Gear Hose Clamp 9/16 W 2 ID PK10
Catalog No. 502768520
Encompass_Preferred
The Worm Gear Hose Clamp 301 Stainless Steel by Oetiker is a high-quality stainless steel worm gear clamp with a 301 stainless steel band. It has a band width of 7/16", a clamping range of 5/8" to 1-1/4", and a torque of 35 in-lbs. This heavy duty hose clamp provides a full 360 degree seal. The 301 stainless steel offers good corrosion resistance. This worm drive clamp is designed for automotive and industrial applications. Overall, this is a durable stainless steel worm gear style hose clamp well-suited for various hoses and fittings.
Catalog No. 50-276-8520 Supplier Grainger Supplier No. 16P257
The Worm Gear Hose Clamp 301 Stainless Steel by Oetiker is a high-quality stainless steel worm gear clamp with a 301 stainless steel band. It has a band width of 7/16", a clamping range of 5/8" to 1-1/4", and a torque of 35 in-lbs. This heavy duty hose clamp provides a full 360 degree seal. The 301 stainless steel offers good corrosion resistance. This worm drive clamp is designed for automotive and industrial applications. Overall, this is a durable stainless steel worm gear style hose clamp well-suited for various hoses and fittings.