Learn More
Grainger Worm Gear Hose Clamp 9/16 W 2.25 ID PK10
Catalog No. 502765140
Encompass_Preferred
The Worm Gear Hose Clamp 301 Stainless Steel by Oetiker is a worm drive hose clamp made from 301 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1/4 inches to 1-1/4 inches. This hose clamp creates a constant radial load around the entire circumference of the hose through the threaded worm drive mechanism. It is designed for automotive and industrial applications. The 301 stainless steel provides good corrosion resistance. This is a high quality hose clamp suitable for demanding environments.
Catalog No. 50-276-5140 Supplier Grainger Supplier No. 16P260
The Worm Gear Hose Clamp 301 Stainless Steel by Oetiker is a worm drive hose clamp made from 301 stainless steel banding. It has a band width of 7/16 inches and a clamping range of 1/4 inches to 1-1/4 inches. This hose clamp creates a constant radial load around the entire circumference of the hose through the threaded worm drive mechanism. It is designed for automotive and industrial applications. The 301 stainless steel provides good corrosion resistance. This is a high quality hose clamp suitable for demanding environments.