Learn More
Grainger Worm Gear Hose Clamp 9/16 W 3/4 ID PK10
Catalog No. 502765938
Encompass_Preferred
The Worm Gear Hose Clamp is a stainless steel worm drive hose clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and is designed for automotive and industrial hose connections. The clamp has a working range of 5/8 inch to 1-1/4 inch diameter. It is made from 201 stainless steel for corrosion resistance. The clamp meets DIN 3017 specifications.
Catalog No. 50-276-5938 Supplier Grainger Supplier No. 16P251
The Worm Gear Hose Clamp is a stainless steel worm drive hose clamp with a band width of 1/2 inch. It has a worm screw drive for tightening and is designed for automotive and industrial hose connections. The clamp has a working range of 5/8 inch to 1-1/4 inch diameter. It is made from 201 stainless steel for corrosion resistance. The clamp meets DIN 3017 specifications.