Learn More
Grainger 5/16" Wide, Interlocked Worm Gear Hose Clamp, 10/pk
Catalog No. NC1002868
Encompass_Preferred
Interlocked worm gear hose clamp helps secure many types of hoses, including pneumatic and exhaust hoses. This general-purpose hose clamp is interlocked to prevent shearing, while clamps used for exhaust have no holes, preventing marring of the hose and fumes escaping. Gear clamp resists rust and corrosion and are useful in humid or damp conditions.10/pk
Catalog No. NC1002868 Supplier Grainger Supplier No. 5X441
Interlocked worm gear hose clamp helps secure many types of hoses, including pneumatic and exhaust hoses. This general-purpose hose clamp is interlocked to prevent shearing, while clamps used for exhaust have no holes, preventing marring of the hose and fumes escaping. Gear clamp resists rust and corrosion and are useful in humid or damp conditions.10/pk