Learn More
NEOGEN Wort Agar, 500 g
SDP
Catalog No. 502015783
Encompass_Preferred
Wort Agar is for the enumeration of yeasts and molds in butter, and is not intended for use in the diagnosis of disease or other conditions in humans. This medium can be modified to enable it to isolate osmophilic yeasts from soft drinks and sugar products by the addition of high concentrations of sucrose and glucose.For more information visit https://www.neogen.com/solutions/Microbiology/wort-agar
Catalog No. 50-201-5783 Supplier NEOGEN Supplier No. 700004662
Wort Agar is for the enumeration of yeasts and molds in butter, and is not intended for use in the diagnosis of disease or other conditions in humans. This medium can be modified to enable it to isolate osmophilic yeasts from soft drinks and sugar products by the addition of high concentrations of sucrose and glucose.For more information visit https://www.neogen.com/solutions/Microbiology/wort-agar