Learn More
Sigma Aldrich Fine Chemicals Biosciences Worthington LD Series Liqu1EA
Catalog No. 502908639
Encompass
The LD Series Liquid Nitrogen Dewars are designed to store and dispense small amounts of liquid nitrogen. Provides high thermal efficiencyRibbed high-strength aluminum body magniformed neck tube design and durable paint make these dewars ruggedEasy to operate design features a lightweight snap-on cap and necktube provides a tight closure and easy access Provides maximum holding times with superior vacuum performance and insulationAccessories sold separately
Catalog No. 50-290-8639 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7439931EA
The LD Series Liquid Nitrogen Dewars are designed to store and dispense small amounts of liquid nitrogen. Provides high thermal efficiencyRibbed high-strength aluminum body magniformed neck tube design and durable paint make these dewars ruggedEasy to operate design features a lightweight snap-on cap and necktube provides a tight closure and easy access Provides maximum holding times with superior vacuum performance and insulationAccessories sold separately